VWR® Traceable®, Ultra-accurate RTD platinum thermometer
Leverandør: VWR Collection
620-1970EA 6150 NOK
620-1970
VWR® Traceable®, Ultra-accurate RTD platinum thermometer
Termometere Digitaltermometer
State-of-the-art aerospace technology delivers high platinum performance.
  • Enhanced precision with platinum probe
  • IP 67 waterproof housing for rugged applications
  • Increase versatility with timer and alarm functions
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
RTD thermometer features a wide –148 to 572 °F and –100 to 300 °C temperature range. The resolution is 0,01 °C across the entire temperature range. This meter features Min/Max memory/HOLD functions and a green and red light HACCP indicator.