Probe for Temperature and Humidity
Leverandør: Testo
0572.6174
620-1973EA 3575 NOK
620-1973
Probe for Temperature and Humidity
Temperaturprober
The thin humidity/temperature probe is suitable for measuring temperature and relative humidity in storerooms and work rooms. The exceptionally small diameter of 4 mm also makes it possible to measure in small holes and narrow passageways.
  • Measures air temperature and humidity quickly and reliably
  • Probe with an exceptionally small diameter of 4 mm
  • Measuring range: humidity 0 to 100% relative humidity; temperature 0 to +40 °C
The thin humidity/temperature probe can be installed for long-term measurements. The probe can be installed using a PG gland orMore Product Information