Single-use temperature loggers
Leverandør: EBRO
1340-6332
620-1974EA 5500 NOK
620-1974
Single-use temperature loggers
Dataloggere
These single-use data loggers are suitable for single-use temperature monitoring during transport of pharmaceutical products, food, medicine and other temperature-sensitive or perishable goods.
  • Configurable high and low limit with visual indication (red LED)
  • Logger is programmable via the free online configuration tool
  • Optionally programmable via FDA 21 CFR Part 11 compliant software: Winlog.basic
  • Eliminates costly, time-consuming IQ/OQ procedures through a standard reporting process
They provide high accuracy and have a convenient USB connectionMore Product Information