Klemme, Labo-Clip
Leverandør: Amarell
K160310
620-1999EA 885 NOK
620-1999
Klemme, Labo-Clip
Termometerholdere
This spring clip holder can be attached quickly for holding thermometers, test tubes and pH meter electrodes. Stainless steel.
  • Rubber base
  • Strudy design
  • Circular support
  • Can be fixed on any container
  • Five holes of different Ø: 4; 5; 6; 10 and 13 mm
Maximum length: 12 mm.