VWR® Traceable®, Digital thermometer, triple display
Leverandør: VWR Collection
620-2032EA 955 NOK
620-2032
VWR® Traceable®, Digital thermometer, triple display
Termometere Digitaltermometer
This digital thermometer is designed for monitoring solutions and room temperatures. It captures and shows the time and date when maximum or minimum values occurred.
  • Accurate sensor and cable, even under water
  • Maximum/minimum (probe) or ambient temperature
  • Wire-mounting bracket for easy probe placement
Internal ambient sensor and external probe sensor (Lר: 38×4,7 mm) with 1,8 metre cable.
Sertifiseringer: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformasjon: Supplied with probe, flip-open stand for the bench, slot forMore Product Information