VWR® Traceable®, Digital thermometer, with data-log memory
Leverandør: VWR Collection
620-2094EA 1125 NOK
620-2094
VWR® Traceable®, Digital thermometer, with data-log memory
Termometere Digitaltermometer
This waterproof thermometer captures 10 temperature readings, stores them in memory, and recalls them on command. For food, agricultural, petrochemical and pharmaceutical applications. The waterproof construction allows use in wet labs, wash-down areas and outside.
  • Maximum/minimum memory readings over any time period
  • 'HOLD' function freezes the display to capture temperature to memory
  • Both high and low audible alarm may be programmed in 1,0 °C increments
  • Stores and recalls 10 temperature readings
Stainless steel probe (L×Ø: 190,5×6,4 mm) with 81,3 cmMore Product Information