VWR® Traceable®, Infrared thermometer
Leverandør: VWR Collection
620-2100EA 2900 NOK
620-2100
VWR® Traceable®, Infrared thermometer
Termometere Digitaltermometer
This infrared thermometer takes instant temperature readings of any surface. It can be used for food, pharmaceuticals, electronics, safety inspection and any liquids or solids. The unique design allows one handed 'point and shoot' readings. Laser-sighting beam enables precision aiming at target surface.
  • Field of view is 11:1
  • Backlighting at the touch of a button
  • Fixed emissivity of 0,95
  • Data "HOLD" function, automatic maximum recording, switchable °C/°F function, low-battery indicator
  • Automatic Off function (after 15 seconds) can be desactivated