VWR® Traceable® excursion trac, Data Logging Thermometers
Leverandør: VWR Collection
620-2204EA 2900 NOK
620-2204 620-2205 620-2206 620-2207 620-2208 620-2209 620-2210
VWR® Traceable® excursion trac, Data Logging Thermometers
Termometere Digitaltermometer
These highly accurate logging thermometers are designed to monitor temperatures in refrigerators, freezers, water baths, heating blocks and incubators.
  • High/low alarms and time stamps
  • User-defined time intervals from 1 min to 24 hours
  • Stores up to 10 unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525 600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data: Unit remains in use while downloading and