Compact data logger
Leverandør: ROTRONIC INSTRUMENTS
LOG-HC2-P1
620-2237EA 7450 NOK
620-2237
Compact data logger
Dataloggere
The compact data logger LOG-HC2-P1 measures and saves humidity and temperature, light, air pressure and acceleration (three-axis) at the same time.
  • High ease of use
  • Four-line LCD with backlight
  • Large memory for more than 2000000 measured values
  • Rechargeable battery for maximum performance
The robust measuring instrument is ideal for long-term data recording.