Multi-use temperature, humidity and shock data logger, 184 G1
Leverandør: Testo
0572.1846
620-2285EA 4500 NOK
620-2285
Multi-use temperature, humidity and shock data logger, 184 G1
Dataloggere
Multi-use temperature, humidity and shock data logger is designed for temperature, humidity and impact monitoring in the cold chain and transportation sectors. It is suitable for the food and pharmaceutical industries for the transport of sensitive goods.
  • Measurement rate from one minute to 24 hours
  • Unlimited operating time
  • Clear indication of alarms and alerts
  • Easy setting and automatic PDF reporting via connection to PC
Ideal for monitoring the transport of sensitive products such as electronics, art, pharmaceuticals or foods safely and conveniently.More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Alarm LED and Display
Display Segmented LCD
Grensesnitt USB
Beskyttelsesklasse IP 30
levetid batteri 120 days
Vekt 44 g
B×D×H 40×97×12 mm
Dataminne - Antall avlesninger 64000
Batterier Lithium battery CR2450, 3 V