VWR® Traceable®, Digital Refrigerator Thermometers
Leverandør: VWR Collection
620-2302EA 370 NOK
620-2302
VWR® Traceable®, Digital Refrigerator Thermometers
Termometere Digitaltermometer
This thermometer is designed for monitoring temperatures in storage rooms, labs, office and fume hoods. It includes attachment devices for flexible positioning.
  • Easy to read LCD
  • Switch from °F to °C
  • Updates every 30 seconds
Sertifiseringer: Traceability provided from ISO/IEC 17025 and standards by NIST.
Leveringsinformasjon: Supplied with suction cup, VELCRO® fastener, silver-oxide battery and Traceable® certificate of calibration.