Food Thermometer, HACCP, TLC 700
Leverandør: EBRO
1340-5735
620-2317EA 855 NOK
620-2317
Food Thermometer, HACCP, TLC 700
Termometere Digitaltermometer
This folding thermometer is suitable for monitoring temperatures quickly and precisely, as required in incoming goods departments, for food distribution, for catering companies and for the food industry and trade, but also for monitoring temperatures in cold-storage warehouses or laboratories. The foldable penetration probe is used to measure the core temperature of food in particular.
  • Robust and shock-resistant
  • Long battery life
  • Small, safe, always with you - for perfect temperature control
  • Two-point factory calibration protocol at –20 and 0 °C