VWR® Traceable® logger trac, Temperature Data Loggers
Leverandør: VWR Collection
620-2322EA 2875 NOK
620-2322 620-2324
VWR® Traceable® logger trac, Temperature Data Loggers
Dataloggere
This temperature logger can display, record and report data during storage, handling and transportation of products.
  • Reusable or re-progammable device with active LCD for cumulative duration of temperature excursion from user-selected minimum/maximum values to be viewed immediately and downloaded if needed
  • LCD shows the running alarm tally based on the acceptable temperature range and recording interval: Alarm tally is incremented when reading goes outside acceptable parameters during recording interval
  • Once an alarm condition has occurred, an 'ALARM'