VWR® Traceable®, Digital barometer
Leverandør: VWR Collection
620-2330EA 3575 NOK
620-2330
VWR® Traceable®, Digital barometer
Barometere
This digital barometer monitors barometric pressure, air temperature, relative humidity also barometric pressure trend and time.
  • Four-line LCD with continuous and simultaneous display of parameters
  • Maximum/minimum readings for pressure, temperature and humidity
  • Data stored every hour for the last 24 hours and can be recalled at the touch of a button
  • Barometric pressure trend shows in bar chart format - current and past 1-, 3-, 6-, 12-, and 24-hour readings
  • Unit can be connected to a computer to permanently capture readings
Sertifiseringer: More Product Information