Multi-use temperature and humidity loggers, testo 184 H1
Leverandør: Testo
0572.1845
620-2361EA 2450 NOK
620-2361
Multi-use temperature and humidity loggers, testo 184 H1
Dataloggere
The 184 H1 temperature and humidity logger is capable of monitoring temperature and humidity levels during cold chain transportation. With configuration, reports, and calibration certificates on-board, the 184 H1 make transport monitoring and documentation of sensitive goods easy.
  • PDF reports allow for quick access to non-corruptible documentation without installed software
  • On-board electronic calibration certificate eliminates the hassles of maintaining traceability
  • Easy configuration on any computer with pre-loaded files, clear alarm indication
  • Its large,