VWR® Durac™ Plus, Hydrometers
Leverandør: VWR Collection
620-2388EA 2700 NOK
620-2388 620-2389
VWR® Durac™ Plus, Hydrometers
Aerometere
These plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
  • Durable and shatterproof, lead free ballast
  • Easy to read black scale on yellow back to avoid eye fatigue
  • Individually serialised - Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Sertifiseringer: NIST.
Bestillingsinformasjon: Includes a multi-language statement of accuracy.
NB: Not recommended for use with petroleum-based liquids or other corrosives, these should be stored in aMore Product Information