pH meters, handheld, PHT 810 / ST 1000
Leverandør: EBRO
1340-5810 1339-0620
EBRO1340-5810EA 3200 NOK
EBRO1340-5810 620-2393
pH meters, handheld, PHT 810 / ST 1000
Elektrokjemiske målere pH-meter
This portable instrument measures the pH value in meat, cold cuts, cheese, dairy products and fluids. It is therefore ideal for butchers, super markets and in the beverage production.
  • Handy and robust
  • Simple calibration with keypad
  • Replaceable electrode
  • Current value memory
  • Fully automatic pH-calibration
  • Battery lifetime approx. 5 years
Leveringsinformasjon: The instrument PHT 810 is supplied without electrode. The set ST 1000 includes the pH meter PHT810, the penetration electrode AT 206, punching pin, buffer solution pH 4/7, protein cleaner, KCl-solution and case.
Bestill nå
SPESIFIKASJONER
Modell ST 1000 PHT 810
Environmentally Preferable
pH - område 0...+14,0 0...+14,0
pH - oppløsning 0,01 0,01
pH - nøyaktighet ±0,03 ±0,03
Temperaturområde −10,0...+50,0 °C −10,0...+50,0 °C
Minne Hold, min. /maks. Hold, min. /maks.
Tilkoblinger BNC BNC
W×D×H 110×54×22 mm 110×54×22 mm
Weight 0,2 kg 0,2 kg