VWR® Traceable®, Digital Thermometer, Platinum, Clinical Trials
Leverandør: VWR Collection
620-2424EA 3575 NOK
620-2424
VWR® Traceable®, Digital Thermometer, Platinum, Clinical Trials
Termometere Digitaltermometer
These platinum clinical trial thermometers are used to monitor the temperature in water baths, heating blocks and incubators. They display min/max temperatures for the previous 24 hours (each hour) and previous seven days (for each days); or previous 31 days (for each day).
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Triple-display simultaneously shows min./max. and current temperatures, °F/°C switchable
  • Micro cable allows refrigerator doors to close on it
  • Alarm is settable in 0,1°