VWR® Traceable®, Digital Thermometer, High Accuracy, Refrigerator and Freezer
Leverandør: VWR Collection
620-2425EA 2650 NOK
620-2425
VWR® Traceable®, Digital Thermometer, High Accuracy, Refrigerator and Freezer
Termometere Digitaltermometer
This high accuracy thermometer monitors both ambient air and the temperature inside a refrigerator or freezer. It reads to 0,01 °C while monitoring MIN/MAX temperatures in the laboratory, and in the refrigerator or freezer simultaneously, with the exact date and time that the readings occurred. It can also be used in water baths, heating blocks and incubators.
  • Smart alarm features a visual/audible alarm that continues to alarm even if the unit returns to non alarm conditions
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is