Temperature data logger starter set, TL-1D
Leverandør: ROTRONIC INSTRUMENTS
TL-1D-SET
620-2465EA 960 NOK
620-2465
Temperature data logger starter set, TL-1D
Dataloggere
This temperature logger is ideal for measuring and recording temperature in food production, storage and transportation. Temperature range: –20 to +70 °C.
  • Compact design with high IP protection
  • Appropriately long battery life (three years at logging interval if 5 min)
  • Programmable start and stop time with the HW4 software
  • Includes HW4-lite programming and analysis software
  • Clear LCD display with visual alerts
  • MIN/MAX/AVG statistical functions via keypad
The values stored are available for download with the included HW4-lite software. After downloadingMore Product Information