VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Leverandør: VWR Collection
620-2486EA 1725 NOK
620-2486 620-2487
VWR® Traceable®, Remote Alarm Thermo-Hygrometer
Hygrometere Digitale termohygrometere
These thermo-hygrometers are ideal for routine measurements, monitoring 'round the clock', all quality control needs, and critical experimental requirements. Perfect for use in hoods, classrooms, stockrooms, environmental chambers and chemical storage areas.
  • Triple-display shows min., max. and current humidity/temperature readings
  • Min./max. temperature and humidity memories
  • High/low alarm feature may be set by user in 1% RH or 1° increments
  • F/°C switchable
  • Remote humidity/temperature sensor on a 2,1 metre cable
* Temperature
** Humidity
***TemperatureMore Product Information