tempmate.®-S1 Single Use Temperature Data Logger
Leverandør: tempmate
S1C10A01000
620-2491EA 5800 NOK
620-2491
tempmate.®-S1 Single Use Temperature Data Logger
Dataloggere
Outstanding single-use temperature logger for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Pre-programmed
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S1 loggers are specially designed for temperature monitoring in any cold chain logistics. As a pre-programmed deviceMore Product Information