Infrared thermometer with circular laserpointer, TFI 260
Leverandør: EBRO
1340-1755
620-2639EA 1050 NOK
620-2639
Infrared thermometer with circular laserpointer, TFI 260
Termometere Digitaltermometer
This basic infrared thermometer measures surface temperatures using infrared technology – within a second and contactless. Therefore, it is ideal for incoming goods inspections, e.g. of foodstuff.
  • Bright background lighting of the display makes it possible to read the measured values even in the dark
  • Antislip handle and trigger for the measurement are optimal for the application
  • Measurement area perfectly marked due to circular laserpointer
  • Distance: spot ratio=12:1
Measuring range: −60 to + 550 °C
Leveringsinformasjon: Calibration certificate included.