Dual-infrared core HACCP thermometer, TLC 750i
Leverandør: EBRO
1340-5736
620-2647EA 2200 NOK
620-2647
Dual-infrared core HACCP thermometer, TLC 750i
Termometere Digitaltermometer
The top-selling TLC 750 infrared thermometer is ideal for non-contact incoming goods inspection with infrared or with the retractable core probe for measuring the core temperature of food. More than 100,000 satisfied customers worldwide!
  • Contamination-free surface measurement and core temperature measurement in one device
  • Surface measurements with infrared; core temperature measurements with penetration probe
  • Display with backlight for reading in dark environments
  • Display can be upside down for reading from both sides
  • Double-laser pointer
  • Watertight