VWR® Traceable®, Infrared Thermometer, with Wristband
Leverandør: VWR Collection
620-2663EA 1675 NOK
620-2663
VWR® Traceable®, Infrared Thermometer, with Wristband
Termometere Digitaltermometer
Traceable® infrared laser thermometer is simple to operate - turn on, point at sample, and take a reading in less than a second. Unit reads both Fahrenheit and Celsius of any surface - solids, semi-solids, and liquids. Non invasive, no touch measurements.
  • Field of view is 6:1
  • Emissivity is 0,05 to 1,00 in 0,01 steps
  • Minimum and maximum feature allows the viewing of highest and lowest readings at any time
  • Laser beam allows perfect sample target sighting
  • Handy wristband
  • Display indicates low battery
Ideal for food preparation, life sciences,More Product Information