VWR®, Weather Station
Leverandør: VWR Collection
620-2692EA 1125 NOK
620-2692
VWR®, Weather Station
Værstasjoner
This weather station is designed for use in agriculture, offices and universities. It includes an internal temperature sensor.
  • Dual-scale and maximum/minimum memory
  • Programmable temperature-alert setting with calendar
  • RF 433 MHz, maximum 30 metre range for remote sensor, wireless thermometer links up to three remote sensors
W×H: 76,2×191 mm
Weight: 363 g
Leveringsinformasjon: Requires 2 AA batteries for main unit and 2 AAA batteries for sensor (included).