Data logger for indoor air quality monitoring, Testo 160 IAQ
Leverandør: Testo
0572.2014
620-2693EA 8750 NOK
620-2693
Data logger for indoor air quality monitoring, Testo 160 IAQ
Dataloggere
We spend a large part of our time indoors – in our homes, workplaces or public buildings. A bad indoor air quality can trigger certain health problems ; mold and fungi growth can cause allergies and asthma ; high CO₂ concentrations can cause headaches and concentration problems. Regular measurement of indoor air quality and comfort levels in workplaces enables the exact setting of the indoor climate and reduces the risk of staff shortages due to illness. Indoor air quality and comfort level measurement therefore also leads to fostering better staff performance.
  • Internal temperature,
More Product Information
Bestill nå
SPESIFIKASJONER
Modell testo 160 IAQ
Environmentally Preferable
Display LCD
Grensesnitt WiFi,USB
Beskyttelsesklasse IP 20
levetid batteri 365 days
Vekt 269 g
B×D×H 117×82×32 mm
Dataminne - Antall avlesninger 32000 measuring values / sum of all channels