CO₂ kit with Bluetooth®, Testo 440
Leverandør: Testo
0563.4405
620-2700EA 10825 NOK
620-2700
CO₂ kit with Bluetooth®, Testo 440
Dataloggere
The CO₂ probe with Bluetooth® is equipped with clearly structured measurement menu for long-term measurement and parallel determination of CO₂ concentration, humidity and air temperature in indoor areas.
  • Mainly used for evaluating the indoor air quality
  • Precise measurement
  • Convenient wireless probe, internal data storage and USB port for data export
Absolute pressure compensation, calculation of wet bulb temperature, dew point and absolute humidity. The air velocity and IAQ measuring instrument can be expanded with a large portfolio of digital probes.