VWR®, Triple-Display Thermometer with Alarm
Leverandør: VWR Collection
620-2716EA 950 NOK
620-2716 620-2717
VWR®, Triple-Display Thermometer with Alarm
Termometere Digitaltermometer
This digital alarm thermometer is for use in ambient conditions. It is designed to control the temperature in cooling and freezing areas in laboratories and drug stores.
  • Optical and acoustic alarm
  • Shows current and max./min. values at the same time
  • Switches between °C and °F
  • 3-point calibration certificate
In addition to the standard alarm function, the acoustic alarm can be confirmed by pushing the Alarm Confirmation button while an acoustic alarm is active. The optical alarm stays until the measured value is back between the limits.