VWR® Traceable®, Data Logging Ethernet Thermometers Compatible with TraceableLIVE® Cloud Service
Leverandør: VWR Collection
620-2737EA 5350 NOK
620-2737 620-2738
VWR® Traceable®, Data Logging Ethernet Thermometers Compatible with TraceableLIVE® Cloud Service
Dataloggere
These data logging thermometers with Ethernet offer an easy and secure way to connect your smartphone, tablet or personal computer remotely with your water baths, refrigerators, freezers and biological/pharmaceutical lab equipment.
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE cloud service
  • Get the reliability, speed, and security using Ethernet connection
  • View current temperature conditions anywhere - cloud-based interface sends email, text, and push notifications
  • No software required
  • Get accurate, reliable measurements with