VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 122 mm Probe
Leverandør: VWR Collection
620-2813EA 610 NOK
620-2813
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 122 mm Probe
Termometere Digitaltermometer
Surface probe stem thermometer is ideal for cooking, laboratories and hot plates.
  • Stainless-steel 120 mm (4,7") probe keep hand away from hot surfaces
  • Will read temperatures in liquids, semi-solids and gases
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Sertifiseringer: Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Leveringsinformasjon: More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Temperaturoppløsning 0,1 °C
Temperaturnøyakighet ±0,5 °C (–10...+100 °C); ±0,9 °F (14...212 °F); otherwise ±1,5 °C / ±2,7 °F
Probe 122 mm (4,8") stainless steel
Kalibreringspunkter 0, 100 °C
Batteritype CR1632
Beskyttelsesklasse Water resistant IP 66
L×B 205×22,5 mm