tempmate.® M2H Multi-Use Temperature and Humidity Data Logger
Leverandør: tempmate
M2H-000
620-2824EA 660 NOK
620-2824
tempmate.® M2H Multi-Use Temperature and Humidity Data Logger
Dataloggere
Intuitive multi-use temperature and humidity logger for reliable low-cost cold chain monitoring. For temperature and humidity recording with internal or external temperature and humidity sensor, multifunction display and automatic PDF report generation.
  • Product-related temperature monitoring along the entire cold chain, in warehouses or fridges
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • Free available software for individual logger configuration
  • Detailed PDF report including graph and table
  • Multifunction