Termometer, innstikk
Leverandør: Testo
0900.0530
620-3123EA 745 NOK
620-3123
Termometer, innstikk
Termometere Digitaltermometer
Small and handy mini thermometer with min./max. alarm.
  • Adjustable min./max. alarm: Acoustic signal sounds when the temperature exceeds or falls below the individually adjustable limit values
  • Permanently connected probe (penetration probe with 80 cm cable)
  • Probe and cable can be stored easily on the temperature measuring instrument
  • With clip for set-up, wall mounting or attachment
The mini alarm thermometer is suitable for temperature measurement in air, in soft powdery substances and in liquids. Using the clips supplied, the mini alarm thermometer canMore Product Information