VWR®, Precision barometer
Leverandør: VWR Collection
628-0041EA 2975 NOK
628-0041
VWR®, Precision barometer
Barometere
This barometer with triple-scale provides a sensitive measure of atmospheric pressure. Corrosion-proof, nickel-chrome.
  • Three distinct scales
  • Adjustable pointer to indicate changes
  • Response time to change from 1 to 4 hours
Sertifiseringer: Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformasjon:
Supplied with altitude adjustment screw (use at locations as high as 1000 metres), wall mount and Traceable® certificate of calibration.