Digital mini thermometer with extended penetration probe
Leverandør: Testo
0560.1111
628-0057EA 550 NOK
628-0057
Digital mini thermometer with extended penetration probe
Termometere Digitaltermometer
This mini thermometer with extended penetration probe shaft is the ideal instrument for taking fast, accurate core temperatures.
  • Digital display shows the exact measured value in °C or °F
  • No reading errors
  • Battery status indicated on display
  • Easy to use and portable in size
Regardless of the field of use, it shows temperatures of up to +250 °C quickly and reliably. The 213 mm long penetration probe allows easy immersion in liquids or penetration of semi-solid substances such as food quality controls, air temperature measurements in HVAC trade.