Digital microscope, DM-1802
Leverandør: MOTIC
DD99421201
630-0552EA 13125 NOK
630-0552
Digital microscope, DM-1802
Mikroskop
This upright microscope is designed for use in education and research projects.
  • Excellent optical quality achieved through innovative design
  • LED illumination with rechargeable batteries for 50 hours of continuous use
  • For advanced teaching thanks to camera and USB interface
Bestillingsinformasjon: Supplied with dust cover.