VWR®, Printer for VWR® Balances
Leverandør: VWR Collection
630-1485EA 6000 NOK
630-1485
VWR®, Printer for VWR® Balances
Printere Document Printers
A compact line thermal dot printer which can be used with VWR® balances.
  • Includes an internal clock
  • Prints date and time together with weight values
  • Print speed 50 mm/s
Leveringsinformasjon: Supplied with connection cable.