Counting chambers
Leverandør: MARIENFELD
0640410 0640510 0640030
630-1510EA 1050 NOK
630-1510 630-1511 630-1509
Counting chambers
Objektglass Tellekammer
These counting chambers are suitable for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
  • Different types of design available
Sertifiseringer: IVD 98/79/EC.
Leveringsinformasjon: Supplied with two cover glasses of 0,4 mm thickness.