Counting chambers
Leverandør: Kova International
87144E 87146E 87100E 87118E 87157E
630-1506EA 3075 NOK
630-1506 630-1539 630-1537 630-1538 630-2048
Counting chambers
Objektglass Tellekammer
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
  • High optical quality through the composition acrylate
  • Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
  • Capillary action for reproducible precision
  • Homogeneous suspension of sample sediments: Retained volume of 1 µl
  • For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl