Counting chamber, Fast Read® 102
Leverandør: Kova International
BVS100H
630-1893EA 1650 NOK
630-1893
Counting chamber, Fast Read® 102
Objektglass Tellekammer
Fast Read 102 systems are made of a disposable plastic slide and designed for the standardisation of microscopic urinalysis.
  • Protected by an optically transparent film
  • Precision and reproducibility guarantee constant readings
  • Identification of the sample by using the numbers printed on the sides
Each chamber is fitted with its own system for the collection of excess urine to prevent any possible contamination.
Chamber depth: 0,1 mm
Chamber volume: 7 µl