VWR® VisiScope® T384 FL, Fluorescence Microscope
Leverandør: VWR Collection
630-1937EA 50625 NOK
630-1937
VWR® VisiScope® T384 FL, Fluorescence Microscope
Mikroskop
Upright microscope for brightfield and fluorescence observations with IOS N-plan objectives. Ideal for routine laboratory applications.
  • Transmitted light (LED) with manual brightness control
  • Double-layer mechanical sliding stage with belt drive in X direction
  • Objectives with anti-fungus treatment
  • Die-cast frame with high stability and ergonomics
Leveringsinformasjon: Supplied ready to use with eyepieces, objectives, dust cover, multi-plug power supply and blue and green filter.