LED illuminated stand magnifier, visolux+
Leverandør: ESCHENBACH OPTIK
1566
630-2181EA 5375 NOK
630-2181
LED illuminated stand magnifier, visolux+
Forstørrelsesglass
This magnifier offers bright and glare-free illumination provided by two SMD LEDs. An integrated pre-selectable orange filter can be used for a warm colour temperature.
  • Automatic light off after 30 minutes
  • Meniscus/aspheric lens for a larger viewing distance
  • Cera-tec® anti-scratch hard coating: Reliability, comfort and relaxed vision
Leveringsinformasjon: Supplied with a hard foam case and three AA batteries.