Kalde lyskilde, KL 1500 HAL
Leverandør: SCHOTT AG Lighting and imaging
150700
630-2597EA 10775 NOK
630-2597
Kalde lyskilde, KL 1500 HAL
Lys Kaldlyskilder
This professional halogen cold light source, for eco-friendly PURAVIS® glass fibre optic light guides, has been developed with the full 150 W halogen spectrum CRI 100, offering the best performance when it comes to colour-critical measurements in life science and industrial applications.
  • Powerful 600 lm light flux
  • Switchable optics ensures uniform, high intensity illumination, even when using light guides with a small diameter
  • Sensor interlock at the light guide socket
  • Ripple-free illumination, perfect for digital imaging applications
  • Ultra-low noise fan