Handheld digital microscope, Dino-Lite Medical, IriScope
Leverandør: Dino-Lite / IDCP B.V.
MEDL4R
630-2615EA 8075 NOK
630-2615
Handheld digital microscope, Dino-Lite Medical, IriScope
Mikroskop
The IriScope enables users to make razor-sharp images of the iris.
  • Handy, easy to use
  • Magnification of 10 to 20 times allows users to see details that can hardly be seen by the naked eye
  • Specially designed front cap can be placed on the eye socket to get the right distance for a perfect picture
  • Built-in LEDs (white + yellow) provide better images of dark irises
  • High speed USB 2.0 output
Sertifiseringer: Medical Device Class 1 – according to Directive 93/42/EEC modified 2007/47/EC.
Leveringsinformasjon: Supplied with protecting pouch, software CD and calibration target.