VWR® VisiCam® TC 20 Plus, Camera with Tablet, Wireless
Leverandør: VWR Collection
630-2661EA 12550 NOK
630-2661
VWR® VisiCam® TC 20 Plus, Camera with Tablet, Wireless
Kamera
This system is composed of a tablet and a wireless 5 MP camera. It can be attached to all microscopes with a C-mount.
  • No need for a computer
  • Wireless connection between tablet and camera: Tablet can be used in any position
  • Images can be shared immediately through wireless network
  • Tablet with user friendly 10,1" touch screen display
  • Quick and easy creation of data reports
Image analysis software pre-installed on the tablet has been developed so that user experience is easy and intuitive. The software allows preview, image capture, video recording,More Product Information
Bestill nå
SPESIFIKASJONER
Modell TC 20 Plus (camera)
Environmentally Preferable
Sensor CMOS
Sensor size 1/2,5"
Piksler (B × H) 1280×800
Bildefrekvens 5 fps
Display High definition LCD (10,1'')
Grensesnitt USB 2.0, WiFi
Programvare VisiCam Image analyser (included)
Supported OS Android
Strømforsyning Via USB