VWR® VisiScope® IT405 FLD, Inverted Fluorescence Microscope
Leverandør: VWR Collection
630-2695EA 110950 NOK
630-2695
VWR® VisiScope® IT405 FLD, Inverted Fluorescence Microscope
Mikroskop
Routine inverted fluorescence microscopes for transmitted brightfield, phase contrast and fluorescence observations with IOS LWD W-Plan PH objectives. Ideal for tissue culture research in clinical and biotechnology laboratories.
  • No warm-up or cool-down times for 60 minute time saving
  • High efficiency LED illumination with brightness control
  • Condenser can be removed to extend the working distance up to 150 mm
  • Fixed specimen stage for fatigue-free operation
  • Two stage insert (glass and metal) with hole for small dimension specimens
  • Plan-achromatic LWD W-Plan