Preparation table for slides
Leverandør: MARIENFELD
6000007
630-2771EA 4175 NOK
630-2771
Preparation table for slides
Objektglassholdere
Preparation table for 20 microscopy slides. Thanks to its recessed grips the slides can be easily taken out.