Desktop lamps, visionLED
Leverandør: ESCHENBACH OPTIK
16036
630-2926EA 2800 NOK
630-2926
Desktop lamps, visionLED
Lys LED-belysnings
High quality desktop lamp offering even, glare-free illumination with high black-white contrast and good colour reproduction.
  • Multi-layer diffuser technology for glare- and hard shadow-free illumination
  • 100% UVA/UVB light blocking
  • Up to 90% blue light damping
  • Colour reproduction index (CRI) of 90
  • LED lifespan: 50000 hours
  • Energy consumption: 10 W
Leveringsinformasjon: Choice of Economy model with 3 colour tones or Comfort model with 5 colour tones and automatic switch off after 2 hours.