VWR® VisiScope® STB150, Stereo Microscope
Leverandør: VWR Collection
630-2942EA 3350 NOK
630-2942
VWR® VisiScope® STB150, Stereo Microscope
Mikroskop
Binocular stereo microscope equipped with turnable objective (2× to 4×), fixed arm with handle, and bright, uniform LED illumination (transmitted and incident). Designed for educational purposes.
  • Adjustable dioptric compensation on both eyepiece tubes
  • Interpupillary distance adjustable between 53 and 75 mm
  • NiMH rechargeable battery for up to 8 hours of use
  • Efficient LED energy saving lamp
Bestillingsinformasjon: Rechargeable batteries not included.
Leveringsinformasjon: Supplied ready to use with eyepieces, objective and dust cover.