VWR® VisiScope® 260, Stereo Microscopes
Leverandør: VWR Collection
630-3072EA 4600 NOK
630-3072 630-3073 630-3074
VWR® VisiScope® 260, Stereo Microscopes
Mikroskop
The cordless VisiScope 260 series of microscopes include binocular stereo microscope, binocular stereo zoom and trinocular stereo zoom versions. Ideal for higher education and laboratory applications.
  • Brightfield observation method for transmitted and incident light
  • Greenough optical system
  • 360° rotating head
  • Long lasting EcoLED illumination
  • Uses Ni-MH rechargeable batteries for up to 12 hours of working time
The stereo zoom models feature a parfocal achromatic zoom.
Bestillingsinformasjon: Rechargeable batteries not included.